missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1 Antibody (XLA1-3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50188FR
This item is not returnable.
View return policy
Description
Cyclin A1 Monoclonal specifically detects Cyclin A1 in Xenopus samples. It is validated for Western Blot.
Specifications
| Cyclin A1 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CCNA1 | |
| Xenopus Cyclin A1 | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 8900 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| XLA1-3 | |
| Western Blot | |
| cyclin A1, cyclin-A1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction