missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A2 Antibody (46B11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2615FR
This item is not returnable.
View return policy
Description
Cyclin A2 Monoclonal specifically detects Cyclin A2 in Xenopus samples. It is validated for Western Blot.
Specifications
| Cyclin A2 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CCNA2 | |
| SDS derived male Xenopus laevis cyclin A2 fusion protein. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 890 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 46B11 | |
| Western Blot | |
| CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction