missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A2 Antibody (E67), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34613DL594
This item is not returnable.
View return policy
Description
Cyclin A2 Monoclonal antibody specifically detects Cyclin A2 in Human, Mouse, Bovine, Mink, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Cyclin A2 | |
| Monoclonal | |
| DyLight 594 | |
| CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 | |
| Full length bovine Cyclin A2 protein (Uniprot: P20248) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 890 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| E67 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine, Mink, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction