missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody (2061A), Alexa Fluor 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB6000AF350
This item is not returnable.
View return policy
Description
Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Cyclin B1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCNB1 | |
| Partial recombinant human Cyclin-B1 protein (amino acids 1-91) [UniProt P14635] | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 2061A | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| Rabbit | |
| 0.1 mL | |
| 891 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction