missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody (2061D), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB60001A
This item is not returnable.
View return policy
Description
Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining)
Specifications
| Cyclin B1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining) | |
| 2061D | |
| Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| E. coli-derived recombinant human Cyclin B1, Met1-Pro91, Accession # P14635 | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors | |
| 891 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction