missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody (SPM619), PE , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11598PE
This item is not returnable.
View return policy
Description
Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| Cyclin B1 | |
| Monoclonal | |
| PE | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| Recombinant human full-length Cyclin B1 protein (Uniprot: P14635) | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors | |
| 891 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| SPM619 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction