missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584499
This item is not returnable.
View return policy
Description
Immunogen sequence: TRNSKINAEN KAKINMAGAK RVPTAPAATS KPGLRPRTAL GDIGNKVSEQ LQAKMPMKKE AKPSATGKVI DK Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 72%.
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.
Specifications
| Cyclin B1 | |
| Polyclonal | |
| Unconjugated | |
| Ccnb1 | |
| Ccn-2; CCNB; Ccnb1; Ccnb1-rs1; Ccnb1-rs13; Cycb; Cycb1; Cycb1-rs1; Cycb-4; Cycb-5; cyclin B; cyclin B1; cyclin B1, related sequence 1; cyclin B1, related sequence 13; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1; I79_013589; OTTHUMP00000221981; OTTHUMP00000223023 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14635 | |
| Ccnb1 | |
| Recombinant protein corresponding to Human CCNB1. Recombinant protein control fragment (Product #RP-105188). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction