Learn More
Cyclin B1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-28603-20UL
 View more versions of this product
Product Code. 18009835
  • € 145.00 / 20µL
Description
The Cyclin B1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Cyclin B1. This antibody recognizes human antigen. The Cyclin B1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Cyclin B1
1000 μg/mL
CCNB, CCNB1, cyclin B1, G2/mitotic specific cyclin B1
20 μL
891
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa