Learn More
Cyclin B2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98962MFV450
This item is not returnable.
View return policy
Description
Cyclin B2 Polyclonal antibody specifically detects Cyclin B2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Immunocytochemistry | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Store at 4C in the dark. |
| 50mM Sodium Borate | |
| 9133 | |
| Purified |