Learn More
Cyclin B2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98962MFV450
Product Code. 18549981
  • € 428.00 / 0.10mL
Description
Cyclin B2 Polyclonal antibody specifically detects Cyclin B2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry
cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299
Store at 4C in the dark.
50mM Sodium Borate
9133