missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Antibody (X121.10), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2612AF532
This item is not returnable.
View return policy
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Xenopus samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cyclin B2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CCNB2 | |
| Xenopus laevis Cyclin B2 | |
| 0.1 mL | |
| Primary | |
| Xenopus species | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| X121.10 | |
| Western Blot, Immunohistochemistry | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Mouse | |
| Protein A purified | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 9133.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction