missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Antibody (X121.10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00
Specifications
| Antigen | Cyclin B2 |
|---|---|
| Clone | X121.10 |
| Concentration | 0.91 mg/ml |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot |
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
| Cyclin B2 | |
| 0.91 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| CCNB2 | |
| IgG1 | |
| Protein A purified | |
| Cyclin B2 (X121.10). Reacts with Xenopus Cyclin B2 only. |
| X121.10 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Xenopus species | |
| 9133 | |
| Xenopus laevis Cyclin B2 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title