missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Antibody (X121.10), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2612MFV610
This item is not returnable.
View return policy
Description
Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| Cyclin B2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Xenopus species | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| X121.10 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Xenopus laevis Cyclin B2 | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 9133 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction