missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Antibody (X29.2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2649B
This item is not returnable.
View return policy
Description
Cyclin B2 Monoclonal specifically detects Cyclin B2 in Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Cyclin B2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CCNB2 | |
| Xenopus laevis cyclin B2 | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 9133 | |
| Mammalia, Xenopus species | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| X29.2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cross-reacts with Cyclin B1 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction