missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin B2 Recombinant Rabbit Monoclonal Antibody (1L9B9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542886
This item is not returnable.
View return policy
Description
Positive test controls include: Rat testis. Immunogen sequence: MALLRRPTVS SDLENIDTGV NSKVKSHVTI RRTVLEEIGN RVTTRAAQVA KKAQNTKVPV QPTKTTNVNK QLKPTASVKP VQMEKLAPKG PSPTPEDVSM.
Cyclin B2 is a member of the cyclin family, specifically the B-type cyclins. The B-type cyclins, B1 and B2, associate with p34cdc2 and are essential components of the cell cycle regulatory machinery. B1 and B2 differ in their subcellular localization. Cyclin B1 co-localizes with microtubules, whereas cyclin B2 is primarily associated with the Golgi region. Cyclin B2 also binds to transforming growth factor beta RII and thus cyclin B2/cdc2 may play a key role in transforming growth factor beta-mediated cell cycle control.
Specifications
| Cyclin B2 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95067, P30276 | |
| Ccnb2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Cyclin B2 (O95067). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1L9B9 | |
| Unconjugated | |
| Ccnb2 | |
| C-B2; CCNB2; ccnb2.S; Cycb2; cyclin B2; cyclin B2 S homeolog; cyclinb2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2; HsT17299; XELAEV_18020679mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12442, 363088, 9133 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction