Learn More
Cyclin D1 Antibody (CL12328) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21214APC
Product Code. 30301865
  • € 737.00 / 0.10mL
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclin D1
Monoclonal
APC
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31
This antibody was developed using a synthetic peptide derived from P24385, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway
595
Store at 4⪚C in the dark.
IgG1
Immunocytochemistry/Immunofluorescence
CL12328
PBS
Mouse
Protein A purified