missing translation for 'onlineSavingsMsg'
Learn More

Cyclin D1 Antibody (CL12328), Novus Biologicals™

Mouse Monoclonal Antibody

€ 329.00 - € 516.00

Specifications

Antigen Cyclin D1
Clone CL12328
Dilution Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Applications Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18615214
View Documents
Novus Biologicals
NBP3-21214-25ul
25 μg
€ 329.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18601275
View Documents
Novus Biologicals
NBP3-21214-100ul
100 μg
€ 516.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

Cyclin D1
Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Monoclonal
Purified
RUO
Human
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31
This antibody was developed against a Synthetic Peptide of P24385. Actual immunogen sequence is proprietary.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
CL12328
Immunofluorescence
Unconjugated
Mouse
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway
PBS, pH 7.2, 40% glycerol
595
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.