Learn More
Novus Biologicals™ Cyclin D1 Antibody (DCS-6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30975-100ug
 View more versions of this product
Product Code. 30067056
  • € 508.00 / 100µg
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human,Mouse,Orangutan,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclin D1
Monoclonal
Unconjugated
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31
A recombinant Cyclin D1 protein
100 μg
Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway
595
Store at -20°C. Avoid freeze/thaw cycles.
IgG2a
Immunohistochemistry, Immunoprecipitation, Western Blot
DCS-6
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Orangutan, Rat