missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D2 Antibody (CCND2/2620), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08804AF532
This item is not returnable.
View return policy
Description
Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human samples. It is validated for ELISA, Protein Array
Specifications
| Cyclin D2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758 | |
| Recombinant full-length human Cyclin D2 (CCND2) protein (Uniprot: IDP30279) | |
| 100 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway | |
| 894 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Protein Array | |
| CCND2/2620 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction