missing translation for 'onlineSavingsMsg'
Learn More

Cyclin D2 Antibody (CCND2/2620), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08804AF750

Product Code. 18506750

  • € 608.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human samples. It is validated for ELISA, Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin D2
Monoclonal
Alexa Fluor 750
cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758
Recombinant full-length human Cyclin D2 (CCND2) protein (Uniprot: IDP30279)
100 μg
Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway
894
Store at 4°C in the dark.
IgG κ
ELISA, Protein Array
CCND2/2620
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.