Learn More
Novus Biologicals™ Cyclin D2 Antibody (DCS3.1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30976-100ug
This item is not returnable.
View return policy
Description
Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Cyclin D2 | |
| Monoclonal | |
| Unconjugated | |
| cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758 | |
| The immunogen for the production of this mouse monoclonal anti-Cyclin D2 was a his-tagged recombinant full length Cyclin D2 34 kDa protein. | |
| 100 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway | |
| 894 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| DCS3.1 | |
| Phosphate buffered saline | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |