Learn More
Novus Biologicals™ Cyclin D2 Antibody (DCS3.1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30976-100ug
 View more versions of this product
Product Code. 30067164
  • € 508.00 / 100µg
Description
Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclin D2
Monoclonal
Unconjugated
cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758
The immunogen for the production of this mouse monoclonal anti-Cyclin D2 was a his-tagged recombinant full length Cyclin D2 34 kDa protein.
100 μg
Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway
894
Store at -20°C. Avoid freeze/thaw cycles.
IgG2a
Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
DCS3.1
Phosphate buffered saline
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Rat