missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D2 Antibody (OTI5A6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70344PCP
This item is not returnable.
View return policy
Description
Cyclin D2 Monoclonal antibody specifically detects Cyclin D2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Cyclin D2 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI5A6 | |
| Western Blot | |
| cyclin D2, G1/S-specific cyclin D2, G1/S-specific cyclin-D2, KIAK0002, MGC102758 | |
| Full length human recombinant protein of human CCND2 (NP_001750) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Wnt Signaling Pathway | |
| 894 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction