missing translation for 'onlineSavingsMsg'
Learn More

Cyclin D2 Monoclonal Antibody (DCS-3.1), Invitrogen™

Product Code. 17777577
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17777577 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17777577 Supplier Invitrogen Supplier No. MA512712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-12712 targets Cyclin D2 in FACS, IF, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-12712 immunogen is purified human recombinant full length cyclin D2 protein.

Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin D2
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DCS-3.1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CCND2
Gene Accession No. P30279, P30280, Q04827
Gene Alias 2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene
Gene Symbols CCND2
Host Species Mouse
Immunogen Purified human recombinant full length cyclin D2 protein
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12444, 64033, 894
Target Species Human, Mouse, Rat
Content And Storage 4° C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.