missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98894-50ul
This item is not returnable.
View return policy
Description
Cyclin D3 Polyclonal antibody specifically detects Cyclin D3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Cyclin D3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 μL/mg of lysate | |
| cyclin D3, D3-type cyclin, G1/S-specific cyclin D3, G1/S-specific cyclin-D3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Cyclin D3. | |
| 50 μg | |
| Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
| 896 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction