missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin E Monoclonal Antibody (HE12), eFluor™ 660, eBioscience™
GREENER_CHOICE

Product Code. 15570107
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15570107 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15570107 Supplier Invitrogen™ Supplier No. 50971480

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: The monoclonal antibody HE12 recognizes human Cyclin E. Cyclins regulate the cell cycle by binding and activating cyclin-dependent kinases (CDK). Cyclin E regulates the transition from G1 to S phase by binding and activating CDK2. The Cyclin E/CDK2 complex is responsible for the phosphorylation of downstream proteins that facilitate the G1 to S phase transition. Overexpression of Cyclin E is found in several types of cancer including breast, colon, bladder, lung and skin. In breast cancer, Cyclin E overexpression is associated with increased stage and grade as well as poor prognosis. Applications Reported: This HE12 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections and microscopy. Applications Tested: This HE12 antibody has been tested by immunohistochemistry of FFPE human tissue using low pH antigen retreival and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Cyclin E belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle. Cyclins function as regulators of CDK kinases. Different cyclins exhibit distinct expression and degradation patterns which contribute to the temporal coordination of each mitotic event. Cyclin E forms a complex with and functions as a regulatory subunit of CDK2, whose activity is required for cell cycle G1/S transition. This protein accumulates at the G1-S phase boundary and is degraded as cells progress through S phase. Overexpression of this gene has been observed in many tumors, which results in chromosome instability, and thus may contribute to tumorigenesis. This protein was found to associate with, and be involved in, the phosphorylation of NPAT protein (nuclear protein mapped to the ATM locus), which participates in cell-cycle regulated histone gene expression and plays a critical role in promoting cell-cycle progression in the absence of pRB. Two alternatively spliced transcript variants of this gene, which encode distinct isoforms, have been described. Two additional splice variants were reported but detailed nucleotide sequence information is not yet available.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin E
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone HE12
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CCNE1
Gene Accession No. P24864
Gene Alias AW538188; Ccne; Ccne1; CycE1; CYCLE; cyclin E; cyclin E1; cyclin Es; cyclin Et; E-S; G1/S-specific cyclin-E1; pCCNE1
Gene Symbols CCNE1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 898
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.