missing translation for 'onlineSavingsMsg'
Learn More

Cyclin E1 Antibody (CCNE1/4935R), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08810AF647

Product Code. 18511563

  • € 669.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin E1
Monoclonal
Alexa Fluor 647
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) (Uniprot: IDP24864)
100 μg
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers
898
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry (Paraffin)
CCNE1/4935R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.