missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E1 Antibody (CCNE1/4935R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08810AF700
This item is not returnable.
View return policy
Description
Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Cyclin E1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
| Recombinant human Cyclin E1 protein fragment (around aa 10-176) (exact sequence is proprietary) (Uniprot: IDP24864) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers | |
| 898 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CCNE1/4935R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction