missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E1, p Thr395 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 479.00
Specifications
| Antigen | Cyclin E1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690227
|
Novus Biologicals
NBP3-05378-100ul |
100 μg |
€ 479.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617276
|
Novus Biologicals
NBP3-05378-20ul |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin E1 Polyclonal antibody specifically detects Cyclin E1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Cyclin E1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 898 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
| A synthetic phosphorylated peptide around T395 of human Cyclin E1 (NP_001229.1). LLTPP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title