missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin E2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-130AF594
This item is not returnable.
View return policy
Description
Cyclin E2 Polyclonal antibody specifically detects Cyclin E2 in Human samples. It is validated for Western Blot
Specifications
| Cyclin E2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| CYCE2, cyclin E2, G1/S-specific cyclin E2, G1/S-specific cyclin-E2 | |
| Partial residue sequence of Cyclin E2 protein. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers | |
| 9134 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction