missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin E2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11548171
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11548171 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11548171 Supplier Invitrogen™ Supplier No. PA14122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-4122 detects Cyclin E2 in human samples. PA1-4122 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~40 kDa protein representing Cyclin E2 in CT10 cell line. The PA1-4122 immunogen is a synthetic peptide corresponding to residues S(2) R R S S R L Q A K Q Q P Q C(15) of Cyclin E2.

This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin E2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CCNE2
Gene Accession No. O96020
Gene Alias Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2
Gene Symbols CCNE2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(2) R R S S R L Q A K Q Q P Q C(15) of Cyclin E2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9134
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.