Learn More
Invitrogen™ Cyclin E2 Recombinant Rabbit Monoclonal Antibody (24GB4750)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557374
This item is not returnable.
View return policy
Description
Cyclin E2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
Specifications
| Cyclin E2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CCNE2 | |
| Ccne2; CYCE2; cyclin E2; G1/S-specific cyclin-E2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9134 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4750 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O96020 | |
| CCNE2 | |
| A synthesized peptide derived from human Cyclin E2 (aa 115-129). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |