Learn More
Invitrogen™ Cyclin G1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592375
This item is not returnable.
View return policy
Description
Immunogen sequence: PSVLALSIIA LEIQAQKCVE LTEGIECLQK HSKINGRDLT FWQELVSKCL TEYSSNKCSK PNVQKLKWIV SGRTARQLKH SYYRITHLPT IPEMVP; Positive Samples: Mouse kidney, Mouse skeletal muscle; Cellular Location: Nucleus.
Cyclins are the regulatory subunits of Cdc2 p34 and related cyclin-dependent kinases (Cdks) which play critical roles in the control of cell cycle progression. The catalytic subunit for cyclin A and B is Cdc2 p34 kinase. The Cdc2- cyclin B complex controls the G2 to M transition whereas Cdc2-cyclin A regulates S phase progression. The G1 to S transition, however, appears to be controlled by the G1 cyclins. Cyclin D1 accumulates during G1 and associates with Cdk2, Cdk4 and Cdk5. Cyclin E and Cdk2 interact during the G1 to S transition. Cyclin G contains a typical N terminal cyclin box and a carboxy terminal domain sequence homologous to the tyrosine phosphorylation site of the epidermal growth factor receptor. Cyclin G expression is induced within 3 hours after growth stimulation and remains elevated with no apparent cell cycle dependency. Cyclin G2 shares 53% amino acid sequence identity with cyclin G1. Peak expression of cyclin G2 is seen in late S phase, as opposed to cyclin G1 expression, which is constitutive.
Specifications
| Cyclin G1 | |
| Polyclonal | |
| Unconjugated | |
| Ccng1 | |
| AI314029; CCNG; CCNG1; CYCG; CYCG1; cyclin G1; cyclin-G; Cyclin-G1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12450, 25405, 900 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P39950, P51945, P51959 | |
| Ccng1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-295 of human Cyclin G1 (NP_0040511). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |