Learn More
Invitrogen™ Cyclin L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540668
This item is not returnable.
View return policy
Description
Peptide sequence: TTTTTTGGIL IGDRLYSEVS LTIDHSLIPE ERLSPTPSMQ DGLDLPSETD Sequence homology: Cow: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.
FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintenance of healthy cells.
Specifications
| Cyclin L1 | |
| Polyclonal | |
| Unconjugated | |
| Ccnl1 | |
| 2610030E23Rik; ANIA6A; ania-6a; AU018493; BM-001; Ccn1; Ccnl; CCNL1; Cyclin Ania-6a; cyclin L ania-6a; cyclin L gamma; cyclin L1; cyclin-L; Cyclin-L1; LOW QUALITY PROTEIN: cyclin-L1; PRO1073; UNQ530/PRO1073 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 57018 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UK58 | |
| Ccnl1 | |
| Synthetic peptide directed towards the N-terminal of human CCNL1 (aa 32-81). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |