missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549750
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Cyclin L1 protein.
FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintenance of healthy cells.
Specifications
| Cyclin L1 | |
| Polyclonal | |
| Unconjugated | |
| Ccnl1 | |
| 2610030E23Rik; ANIA6A; ania-6a; AU018493; BM-001; Ccn1; Ccnl; CCNL1; Cyclin Ania-6a; cyclin L ania-6a; cyclin L gamma; cyclin L1; cyclin-L; Cyclin-L1; LOW QUALITY PROTEIN: cyclin-L1; PRO1073; UNQ530/PRO1073 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114121, 56706, 57018 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q52KE7, Q9R1Q2, Q9UK58 | |
| Ccnl1 | |
| Synthesized peptide derived from human Cyclin-L1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction