missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin M3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32134JF646
This item is not returnable.
View return policy
Description
Cyclin M3 Polyclonal antibody specifically detects Cyclin M3 in Human samples. It is validated for Western Blot
Specifications
| Cyclin M3 | |
| Polyclonal | |
| Western Blot | |
| ACDP3cyclin-M3, ancient conserved domain protein 3, Ancient conserved domain-containing protein 3, cyclin M3, Cyclin-M3, DKFZp434I1016, FLJ20018, metal transporter CNNM3 | |
| The immunogen this antibody was made to, maps to a region between residue 657 to 707 of human Cyclin M3 using the numbering given in entry NP_060093.3 (GeneID 26505). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 26505 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction