missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin M3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32133
This item is not returnable.
View return policy
Description
Cyclin M3 Polyclonal specifically detects Cyclin M3 in Human samples. It is validated for Immunoprecipitation.
Specifications
| Cyclin M3 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| ACDP3cyclin-M3, ancient conserved domain protein 3, Ancient conserved domain-containing protein 3, cyclin M3, Cyclin-M3, DKFZp434I1016, FLJ20018, metal transporter CNNM3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26505 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2 - 10 mcg/mg lysate | |
| NP_060093.3 | |
| CNNM3 | |
| The immunogen this antibody was made to, maps to a region between residue 575 to 625 of human Cyclin M3 using the numbering given in entry NP_060093.3 (GeneID 26505). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction