missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin T1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81898R
This item is not returnable.
View return policy
Description
Cyclin T1 Polyclonal antibody specifically detects Cyclin T1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cyclin T1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| CCNT, CCNT1, CCNTcyclin-T, CDK9-associated C-type protein, cyclin C-related protein, cyclin T1, cyclin T1b, Cyclin-T, cyclin-T1, CYCT1, HIVE1, human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1, Human immunodeficiency virus-1 expression, subunit of positive elongation transcription factor b | |
| Rabbit polyclonal Cyclin T1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human Cyclin T1. The immunogen is located within the last 50 amino acids of Cyclin T1. Amino Acid Squence: NTDKKPRPPPLPSEPPPPL | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 904 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction