Learn More
Cyclododecanone, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Cyclododecanone |
| CAS | 830-13-7 |
| Molecular Formula | C12H22O |
| Quantity | 5 g |
| Synonym | NSC 77116 |
| InChI Formula | InChI=1S/C12H22O/c13-12-10-8-6-4-2-1-3-5-7-9-11-12/h1-11H2 |
| SMILES | O=C1CCCCCCCCCCC1 |
| IUPAC Name | cyclododecanone |
| Molecular Weight (g/mol) | 182.3 |
| Formula Weight | 182.1671 |