missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexyl Butyl Phthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Cyclohexyl Butyl Phthalate-d4 |
| Molecular Formula | C18 D4 H20 O4 |
| Quantity | 5 mg |
| Synonym | 1-Butyl 2-cyclohexyl Ester 1,2-Benzenedicarboxylic Acid-d4, Butyl Cyclohexyl Ester1,2-Benzenedicarboxylic Acid-d4, Phthalic Acid-d4, Butyl Cyclohexyl Ester-d4, Butyl Cyclohexyl Phthalate-d4, Elastex 50B-d4, NSC 69994-d4 |
| InChI Formula | InChI=1S/C18H24O4/c1-2-3-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-9-5-4-6-10-14/h7-8,11-12,14H,2-6,9-10,13H2,1H3/i7D,8D,11D,12D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OC2CCCCC2)c(C(=O)OCCCC)c1[2H] |
| IUPAC Name | 1-O-butyl 2-O-cyclohexyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 308.405 |
| Formula Weight | 308.193 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?