missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentolate Hydrochloride, TRC

Product Code. 30135023
Change view
Click to view available options
Quantity:
25 mg
50 mg
500 mg
Unit Size:
25mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135023 50 mg 50mg
30133877 500 mg 500mg
30011229 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135023 Supplier Toronto Research Chemicals Supplier No. TRCC98843050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5870-29-1
Molecular Formula C17 H25 N O3 . Cl H
Molecular Weight (g/mol) 327.85
Synonym Cyclopentolate Hydrochloride, 2-(Dimethylamino)ethyl (2RS)-(1-hydroxycyclopentyl)(phenyl)acetate hydrochloride
IUPAC Name 2-(dimethylamino)ethyl 2-(1-hydroxycyclopentyl)-2-phenylacetate;hydrochloride
SMILES Cl.CN(C)CCOC(=O)C(c1ccccc1)C2(O)CCCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H25NO3.ClH/c1-18(2)12-13-21-16(19)15(14-8-4-3-5-9-14)17(20)10-6-7-11-17;/h3-5,8-9,15,20H,6-7,10-13H2,1-2H3;1H
Formula Weight 327.1601
Chemical Name or Material Cyclopentolate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.