missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin 40 Antibody (SR1688), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22135-50ul
This item is not returnable.
View return policy
Description
Cyclophilin 40 Monoclonal antibody specifically detects Cyclophilin 40 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
| Cyclophilin 40 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| SR1688 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| 40 kDa peptidyl-prolyl cis-trans isomerase, 40 kDa peptidyl-prolyl cis-trans isomerase D, cyclophilin D, Cyclophilin-40, Cyclophilin-related protein, CYP40, CYP-40cyclophilin 40, CYPD, EC 5.2.1.8, MGC33096, peptidyl-prolyl cis-trans isomerase D, peptidylprolyl isomerase D, peptidylprolyl isomerase D (cyclophilin D), PPIase D, Rotamase D | |
| A synthesized peptide derived from human Cyclophilin 40 (Uniprot #: Q08752) | |
| 50 μg | |
| Immunology | |
| 5481 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction