missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98906-50ul
This item is not returnable.
View return policy
Description
Cyclophilin B Polyclonal antibody specifically detects Cyclophilin B in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Cyclophilin B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 μL/mg of lysate | |
| Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Cyclophilin B. | |
| 50 μg | |
| Loading Controls | |
| 5479 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction