Learn More
Invitrogen™ Cyclophilin B Monoclonal Antibody (M019)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547662
Description
Clone M019 detects a 20 kDa* band corresponding to the molecular mass of cyclophilin B on SDS-PAGE immunoblots of human LNCaP, MeWo, MCF7, and MDA-MB-231 cell lysates, as well as full length recombinant cyclophilin B, but does not detect recombinant cyclophilin A. The antibody can be used for western blot, immunoprecipitation, protein ELISA, and immunocytochemistry.
Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins which bind FK506 are termed FK506 Binding Proteins (FKBP's) and those which bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation. Thus, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins.
Specifications
| Cyclophilin B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P23284 | |
| PPIB | |
| Clone M019 was generated from a proprietary antigen related to cyclophilin B from the MCF7 breast cancer cell line. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M019 | |
| Unconjugated | |
| PPIB | |
| AA408962; AA553318; AI844835; Cphn2; Cphn-2; cyclophilin B; cyclophilin-like protein; Cyp B; CyP-20b; CYPB; CYP-S1; EC 5.2.1.8; epididymis secretory protein Li 39; HEL-S-39; OI9; Peptidyl-prolyl cis-trans isomerase B; peptidylprolyl isomerase B; peptidylprolyl isomerase B (cyclophilin B); PPIase; PPIase B; Ppib; Rotamase; Rotamase B; S-cyclophilin; S-cyclophylin; SCYLP | |
| Mouse | |
| Protein G | |
| RUO | |
| 5479 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.