missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin-E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | Cyclophilin-E |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661446
|
Novus Biologicals
NBP2-99016-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681296
|
Novus Biologicals
NBP2-99016-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclophilin-E Polyclonal antibody specifically detects Cyclophilin-E in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Cyclophilin-E | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 0.2 μm filtered solution in PBS | |
| 10450 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.5-2 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Cyclophilin-E. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title