missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclophilin E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564978
This item is not returnable.
View return policy
Description
Immunogen sequence: EGSEPPKAETQ EGEPIAKKAR SNPQVYMDIK IGNKPAGRIQ MLLRSDVVPM TAENFRCLCT HEKGFGFKGS Highest antigen sequence identity to the following orthologs - mouse 96%, rat 94%.
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein contains a highly conserved cyclophilin (CYP) domain as well as an RNA-binding domain. It was shown to possess PPIase and protein folding activities and also exhibit RNA-binding activity. Three alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| Cyclophilin E | |
| Polyclonal | |
| Unconjugated | |
| PPIE | |
| 2010010D16Rik; Ab1-210; Cyclophilin E; cyclophilin-33; CYP33; CYP-33; Peptidyl-prolyl cis-trans isomerase E; peptidylprolyl isomerase E; peptidylprolyl isomerase E (cyclophilin E); PPIase E; PPIE; Rotamase E | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UNP9 | |
| PPIE | |
| Recombinant Human Cyclophilin E. Recombinant protein control fragment (Product #RP-105746). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction