missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin-F Antibody (1F5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28608C
This item is not returnable.
View return policy
Description
Cyclophilin-F Monoclonal antibody specifically detects Cyclophilin-F in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cyclophilin-F | |
| Monoclonal | |
| DyLight 650 | |
| P30405 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1F5 | |
| 50mM Sodium Borate | |
| Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F) | |
| Recombinant human Cyclophilin-F (30-207aa) purified from E. coli (NP_005720). | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 10105 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction