missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin-F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38034-20ul
This item is not returnable.
View return policy
Description
Cyclophilin-F Polyclonal antibody specifically detects Cyclophilin-F in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Cyclophilin-F | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-207 of human Cyclophilin-F (NP_005720.1).,, Sequence:, CSKGSGDPSSSSSSGNPLVYLDVDANGKPLGRVVLELKADVVPKTAENFRALCTGEKGFGYKGSTFHRVIPSFMCQAGDFTNHNGTGGKSIYGSRFPDENFTLKHVGPGVLSMANAGPNTNGSQFFICTIKTDWLDGKHVVFGHVKEGMDVVKKIESFGSKSGRTSKKIVITDCGQLS | |
| 20 μL | |
| Proteases & Other Enzymes | |
| 10105 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction