missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin-F Antibody (OTI1G8), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71551C
This item is not returnable.
View return policy
Description
Cyclophilin-F Monoclonal specifically detects Cyclophilin-F in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cyclophilin-F | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PPIF | |
| Full length recombinant protein of human Cyclophilin-F (NP_005720) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1G8 | |
| Western Blot, Immunohistochemistry | |
| Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F) | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 10105.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction