Learn More
Cyclophosphamide Monohydrate, TRC
Product Code. 30012129
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012129 25 g 25g
30133001 1 g 1g
30134730 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012129 Supplier Toronto Research Chemicals Supplier No. TRCC98858025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6055-19-2
Molecular Formula C7 H15 Cl2 N2 O2 P . H2 O
Molecular Weight (g/mol) 279.1
Synonym 2H-1,3,2-Oxazaphosphorin-2-amine, N,N-bis(2-chloroethyl)tetrahydro-, 2-oxide, monohydrate (9CI), 2H-1,3,2-Oxazaphosphorine, 2-[bis(2-chloroethyl)amino]tetrahydro-, 2-oxide, monohydrate (8CI), 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine-2-oxide monohydrate, Cyclophosphamide hydrate, Cyclophosphamide monohydrate, Endoxan monohydrate
IUPAC Name N,N-bis(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine;hydrate
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2
Formula Weight 278.0354
Percent Purity >95%
Chemical Name or Material Cyclophosphamide Monohydrate
Grade HPLC