Learn More
Cyclopropyl(4-fluorophenyl)methanone, TRC
Specifications
Specifications
| Chemical Name or Material | Cyclopropyl(4-fluorophenyl)methanone |
| CAS | 772-31-6 |
| Molecular Formula | C10H9OF |
| Quantity | 10 mg |
| Synonym | Cyclopropyl 4-Fluorophenyl Ketone |
| InChI Formula | InChI=1S/C10H9FO/c11-9-5-3-8(4-6-9)10(12)7-1-2-7/h3-7H,1-2H2 |
| SMILES | Fc1ccc(cc1)C(=O)C2CC2 |
| IUPAC Name | cyclopropyl-(4-fluorophenyl)methanone |
| Molecular Weight (g/mol) | 164.17 |
| Formula Weight | 164.0637 |