Learn More
CYCS Mouse anti-Human, Pigeon, Clone: SPM389, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of CYCS.
Brand: Abnova MAB13139.100ug
Description
This gene encodes cytochrome c, a component of the electron transport chain in mitochondria. The heme group of cytochrome c accepts electrons from the b-c1 complex and transfers electrons to the cytochrome oxidase complex. Cytochrome c is also involved in initiation of apoptosis. Upon release of cytochrome c to the cytoplasm, the protein binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Many cytochrome c pseudogenes exist, scattered throughout the human genome. [provided by RefSeq]
Specifications
| CYCS | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be d | |
| CYC/HCS | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 54205 | |
| Store at 4°C. | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| SPM389 | |
| Mouse monoclonal antibody raised against synthetic peptide of CYCS. | |
| In 1mM PBS (0.05% BSA, 0.05% sodium azide). | |
| CYCS | |
| A synthetic peptide corresponding to amino acids 1-80, 81-104 and 66-104 of pigeon CYCS. | |
| 100 μg | |
| Primary | |
| Human, Pigeon | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.